Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-66
Entrez Gene ID
191824
Gene Name
Serpentine Receptor, class D (delta)
Gene Aliases
CELE_Y39A3B.4
Chromosome Location
Chr.III: 1980823 - 1982647 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065086.2 NP_497487.2 1 - 2 195 70

View Full Product Details