Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cpt-3
Entrez Gene ID
190055
Gene Name
Carn_acyltransf domain-containing protein
Gene Aliases
CELE_Y48G9A.10
Chromosome Location
Chr.III: 2082819 - 2088314 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065097.5 NP_497498.2 1 - 2 100 92

View Full Product Details