Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
frl-1
Entrez Gene ID
175346
Gene Name
FRL (Formin Related gene in Leukocytes) homolog
Gene Aliases
CELE_Y48G9A.4
Chromosome Location
Chr.III: 2124137 - 2151554 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065104.7 NP_497505.3 7 - 8 1294 138

View Full Product Details