Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
fbxa-48
Entrez Gene ID
190273
Gene Name
F-box domain-containing protein
Gene Aliases
CELE_Y54F10BM.7
Chromosome Location
Chr.III: 2266980 - 2272175 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065129.5 NP_497530.2 1 - 2 160 106

View Full Product Details