Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ceh-44
Entrez Gene ID
175372
Gene Name
protein-coding
Gene Aliases
CELE_Y54F10AM.4
Chromosome Location
Chr.III: 2530606 - 2546420 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065175.7 NP_497576.1 7 - 8 1226 106

View Full Product Details