Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
set-27
Entrez Gene ID
190607
Gene Name
SET domain-containing protein
Gene Aliases
CELE_Y71H2AM.8
Chromosome Location
Chr.III: 2700937 - 2707846 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065203.4 NP_497604.2 2 - 3 207 72

View Full Product Details