Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gly-14
Entrez Gene ID
175453
Gene Name
GLYcosylation related
Gene Aliases
CELE_M01F1.1
Chromosome Location
Chr.III: 3490337 - 3493238 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065318.4 NP_497719.1 5 - 6 607 168
GenBank mRNA AF082012.1 - 5 - 6 601 168

View Full Product Details