Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lin-48
Entrez Gene ID
175485
Gene Name
Uncharacterized protein
Gene Aliases
CELE_F34D10.5
Chromosome Location
Chr.III: 3745438 - 3747572 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_065358.5 NP_497759.1 5 - 6 704 124
GenBank mRNA AF134806.1 - 5 - 6 706 124

View Full Product Details