Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wht-7
Entrez Gene ID
175864
Gene Name
protein-coding
Gene Aliases
CELE_Y42G9A.6
Chromosome Location
Chr.III: 6141959 - 6149597 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001379747.1 NP_001367851.1 6 - 7 675 62
NM_001379748.1 NP_001367852.1 2 - 3 252 62

View Full Product Details