Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
srg-4
Entrez Gene ID
24104888
Gene Name
Serpentine receptor class gamma-4
Gene Aliases
CELE_T12A2.12
Chromosome Location
Chr.III: 6259653 - 6262085 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001306703.2 NP_001293632.1 4 - 5 823 105

View Full Product Details