Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
odr-4
Entrez Gene ID
175950
Gene Name
protein-coding
Gene Aliases
CELE_Y102E9.1
Chromosome Location
Chr.III: 6730737 - 6748157 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001027643.4 NP_001022814.1 3 - 4 491 65
NM_001027644.8 NP_001022815.1 3 - 4 442 65
NM_001382900.1 2 - 3 294 65
GenBank mRNA AF055911.1 - 1 - 2 199 65

View Full Product Details