Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
dcr-1
Entrez Gene ID
176138
Gene Name
Death-promoting deoxyribonuclease
Gene Aliases
CELE_K12H4.8
Chromosome Location
Chr.III: 8071967 - 8080281 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066360.4 NP_498761.2 18 - 19 3911 131

View Full Product Details