Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
hlh-4
Entrez Gene ID
176372
Gene Name
Helix-loop-helix protein 4
Gene Aliases
CELE_T05G5.2
Chromosome Location
Chr.III: 9737477 - 9743338 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382969.1 9 - 10 1758 66

View Full Product Details