Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
arl-5
Entrez Gene ID
176392
Gene Name
ADP-ribosylation factor-like protein 5
Gene Aliases
CELE_ZK632.8
Chromosome Location
Chr.III: 9819303 - 9824133 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001382971.1 10 - 11 1898 81

View Full Product Details