Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-47
Entrez Gene ID
176431
Gene Name
Vesicular GABA transporter
Gene Aliases
CELE_T20G5.6
Chromosome Location
Chr.III: 10183757 - 10186260 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066854.6 NP_499255.1 3 - 4 599 82
GenBank mRNA AF031935.1 - 3 - 4 599 82

View Full Product Details