The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_066854.6 | NP_499255.1 | 5 - 6 | 1275 | 109 |
GenBank mRNA | AF031935.1 | - | 5 - 6 | 1275 | 109 |