Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
oxi-1
Entrez Gene ID
176515
Gene Name
HECT domain-containing protein
Gene Aliases
CELE_Y39A1C.2
Chromosome Location
Chr.III: 10858284 - 10867033 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_066991.5 NP_499392.1 4 - 5 1026 65

View Full Product Details