Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
smc-3
Entrez Gene ID
176559
Gene Name
protein-coding
Gene Aliases
CELE_Y47D3A.26
Chromosome Location
Chr.III: 11290911 - 11306247 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268189.3 NP_001255118.1 2 - 3 167 103

View Full Product Details