Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nft-1
Entrez Gene ID
176628
Gene Name
Nitrilase homolog
Gene Aliases
CELE_Y56A3A.13
Chromosome Location
Chr.III: 11923886 - 11929387 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067155.8 NP_499556.1 7 - 8 1173 87
GenBank mRNA AF069986.1 - 7 - 8 1171 87

View Full Product Details