Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
elpc-2
Entrez Gene ID
176687
Gene Name
ELongator complex Protein Component
Gene Aliases
CELE_Y111B2A.17
Chromosome Location
Chr.III: 12669043 - 12686248 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067247.10 NP_499648.3 2 - 3 318 72

View Full Product Details