Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-8
Entrez Gene ID
177494
Gene Name
protein-coding
Gene Aliases
CELE_R13A1.4
Chromosome Location
Chr.IV: 7197153 - 7203660 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307363.3 NP_001294292.1 10 - 11 1041 116
NM_001307364.3 NP_001294293.1 9 - 10 1047 116
NM_001307365.3 NP_001294294.1 8 - 9 924 116
NM_068737.2 NP_501138.1 10 - 11 1044 116
GenBank mRNA U76402.1 - 10 - 11 1044 116

View Full Product Details