Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nol-10
Entrez Gene ID
177538
Gene Name
NUC153 domain-containing protein
Gene Aliases
CELE_F32E10.1
Chromosome Location
Chr.IV: 7580597 - 7583726 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068832.5 NP_501233.1 6 - 7 1880 84

View Full Product Details