Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
lgc-5
Entrez Gene ID
184620
Gene Name
Neur_chan_LBD domain-containing protein
Gene Aliases
CELE_F17E9.7
Chromosome Location
Chr.IV: 8342685 - 8344532 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069010.5 NP_501411.4 3 - 4 357 81

View Full Product Details