Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cka-1
Entrez Gene ID
177807
Gene Name
protein-coding
Gene Aliases
CELE_C28D4.2
Chromosome Location
Chr.IV: 9731737 - 9737621 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136377.4 NP_001129849.1 3 - 4 431 120
NM_001306954.3 NP_001293883.1 3 - 4 433 120
NM_069331.4 NP_501732.1 4 - 5 509 120

View Full Product Details