Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
kin-5
Entrez Gene ID
188489
Gene Name
Tyrosine-protein kinase
Gene Aliases
CELE_T13H10.1
Chromosome Location
Chr.IV: 9967077 - 9969181 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069392.5 NP_501793.1 8 - 9 1537 80

View Full Product Details