Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- gst-4
- Entrez Gene ID
-
177886
- Gene Name
- Glutathione S-transferase 4
- Gene Aliases
-
CELE_K08F4.7
- Chromosome Location
-
Chr.IV: 10142135 - 10143099 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069447.7 |
NP_501848.1 |
1 - 2 |
240 |
84 |
GenBank mRNA |
AF010239.1 |
- |
1 - 2 |
135 |
84 |
- Gene Symbol
- dhs-12
- Entrez Gene ID
-
177888
- Gene Name
- DeHydrogenases, Short chain
- Gene Aliases
-
CELE_K08F4.9
- Chromosome Location
-
Chr.IV: 10142199 - 10146225 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_069449.7 |
NP_501850.1 |
1 - 2 |
176 |
84 |
View Full Product Details