The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001306925.2 | NP_001293854.1 | 21 - 22 | 3182 | 82 |
NM_001306926.3 | NP_001293855.1 | 18 - 19 | 2945 | 82 |