Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
unc-43
Entrez Gene ID
177921
Gene Name
protein-coding
Gene Aliases
CELE_K11E8.1
Chromosome Location
Chr.IV: 10324254 - 10350292 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028122.5 NP_001023293.2 3 - 4 282 69
NM_001028127.6 NP_001023298.1 13 - 14 1457 69
NM_001028133.5 NP_001023304.1 3 - 4 767 69
NM_001129316.4 NP_001122788.1 3 - 4 410 69
NM_001136391.4 NP_001129863.1 3 - 4 240 69
NM_001268551.3 NP_001255480.1 15 - 16 1635 69
NM_001268552.3 NP_001255481.1 4 - 5 390 69
NM_001268553.3 NP_001255482.1 3 - 4 486 69
NM_001380439.2 NP_001366993.1 14 - 15 1603 69
NM_001380440.2 NP_001366994.1 15 - 16 1710 69
NM_001380441.2 NP_001366996.1 9 - 10 1158 69
NM_001383187.1 13 - 14 1464 69
GenBank mRNA AF233262.1 - 11 - 12 1152 69
AF233264.1 - 12 - 13 1305 69
AF233265.1 - 11 - 12 1146 69
AF233266.1 - 9 - 10 1004 69

View Full Product Details