Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tax-6
Entrez Gene ID
177943
Gene Name
protein-coding
Gene Aliases
CELE_C02F4.2
Chromosome Location
Chr.IV: 10492869 - 10501600 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026121.2 NP_001021292.1 11 - 12 1573 68

View Full Product Details