Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- ndx-9
- Entrez Gene ID
-
177996
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F13H10.2
- Chromosome Location
-
Chr.IV: 11009981 - 11012020 on Build WBcel235
- Gene Symbol
- F13H10.3
- Entrez Gene ID
-
177997
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F13H10.3
- Chromosome Location
-
Chr.IV: 11010037 - 11015953 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_171417.7 |
NP_741486.1 |
16 - 17 |
3325 |
79 |
View Full Product Details