Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
rap-3
Entrez Gene ID
182417
Gene Name
RAP homolog (vertebrate Rap GTPase family)
Gene Aliases
CELE_C08F8.7
Chromosome Location
Chr.IV: 11177267 - 11178241 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_069694.4 NP_502095.2 1 - 2 78 86

View Full Product Details