Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
madf-1
Entrez Gene ID
190316
Gene Name
protein-coding
Gene Aliases
CELE_Y55F3BR.5
Chromosome Location
Chr.IV: 830358 - 831896 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307533.3 NP_001294462.1 2 - 3 270 93
NM_001380020.2 NP_001367655.1 3 - 4 312 93

View Full Product Details