Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
plx-1
Entrez Gene ID
246019
Gene Name
protein-coding
Gene Aliases
CELE_Y55F3AL.1
Chromosome Location
Chr.IV: 959521 - 984395 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307625.3 NP_001294554.1 9 - 10 1908 112
NM_001307626.3 NP_001294555.1 6 - 7 1341 112
NM_001307627.3 NP_001294556.1 6 - 7 1341 112
NM_067617.6 NP_500018.3 9 - 10 1908 112

View Full Product Details