Assay Design
The assay probe spans an exon junction.
Gene Transcripts
- Gene Symbol
- cmk-1
- Entrez Gene ID
-
176989
- Gene Name
- Calcium/calmodulin-dependent protein kinase type 1
- Gene Aliases
-
CELE_K07A9.2
- Chromosome Location
-
Chr.IV: 1824481 - 1834247 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_067738.8 |
NP_500139.1 |
2 - 3 |
274 |
105 |
View Full Product Details