Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cmk-1
Entrez Gene ID
176989
Gene Name
Calcium/calmodulin-dependent protein kinase type 1
Gene Aliases
CELE_K07A9.2
Chromosome Location
Chr.IV: 1824481 - 1834247 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067738.8 NP_500139.1 3 - 4 426 68

View Full Product Details