Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
eri-5
Entrez Gene ID
177029
Gene Name
protein-coding
Gene Aliases
CELE_Y38F2AR.1
Chromosome Location
Chr.IV: 2390821 - 2396263 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_067798.6 NP_500199.2 2 - 3 666 98

View Full Product Details