Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pax-2
Entrez Gene ID
187062
Gene Name
protein-coding
Gene Aliases
CELE_K06B9.5
Chromosome Location
Chr.IV: 4198973 - 4204375 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068112.4 NP_500513.1 1 - 2 75 112

View Full Product Details