Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
fem-1
Entrez Gene ID
177335
Gene Name
protein-coding
Gene Aliases
CELE_F35D6.1
Chromosome Location
Chr.IV: 5533140 - 5537992 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068423.6 NP_500824.1 2 - 3 338 74
NM_068424.4 NP_500825.1 3 - 4 297 74

View Full Product Details