Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srv-21
Entrez Gene ID
186686
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_H04M03.8
Chromosome Location
Chr.IV: 5876092 - 5877346 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_068479.1 NP_500880.1 2 - 3 300 150

View Full Product Details