Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-44
Entrez Gene ID
177366
Gene Name
protein-coding
Gene Aliases
CELE_B0350.2
Chromosome Location
Chr.IV: 5975557 - 6004857 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026096.4 NP_001021267.1 4 - 5 2298 68
NM_001380249.1 NP_001367345.1 12 - 13 5358 68
NM_001380250.2 NP_001367343.1 11 - 12 5359 68
NM_001380252.2 NP_001368407.1 12 - 13 5363 68
NM_001380253.1 NP_001368408.1 6 - 7 2841 68
NM_001380254.1 NP_001368409.1 7 - 8 2859 68
NM_068500.5 NP_500901.2 5 - 6 2628 68

View Full Product Details