Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-44
Entrez Gene ID
177366
Gene Name
protein-coding
Gene Aliases
CELE_B0350.2
Chromosome Location
Chr.IV: 5975557 - 6004857 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380249.1 NP_001367345.1 3 - 4 1607 99
NM_001380250.2 NP_001367343.1 3 - 4 1626 99
NM_001380251.2 NP_001367344.1 3 - 4 1623 99
NM_001380252.2 NP_001368407.1 3 - 4 1612 99

View Full Product Details