Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-44
Entrez Gene ID
177366
Gene Name
protein-coding
Gene Aliases
CELE_B0350.2
Chromosome Location
Chr.IV: 5975557 - 6004857 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001380249.1 NP_001367345.1 17 - 18 20816 78

View Full Product Details