Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
pqn-89
Entrez Gene ID
177412
Gene Name
protein-coding
Gene Aliases
CELE_Y73B6BR.1
Chromosome Location
Chr.IV: 6506724 - 6511113 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028406.5 NP_001023577.1 2 - 3 337 114
NM_001383105.1 2 - 3 339 114

View Full Product Details