Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nhr-43
Entrez Gene ID
178121
Gene Name
protein-coding
Gene Aliases
CELE_C29E6.5
Chromosome Location
Chr.IV: 11899590 - 11902976 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001307047.3 NP_001293976.1 7 - 8 1415 125
NM_069853.5 NP_502254.1 7 - 8 1350 125

View Full Product Details