Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
unc-30
Entrez Gene ID
178265
Gene Name
protein-coding
Gene Aliases
CELE_B0564.10
Chromosome Location
Chr.IV: 13118977 - 13124406 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001026105.4 NP_001021276.1 6 - 7 992 63
NM_001268772.4 NP_001255701.1 6 - 7 724 63

View Full Product Details