Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
egl-23
Entrez Gene ID
178358
Gene Name
protein-coding
Gene Aliases
CELE_Y37A1B.11
Chromosome Location
Chr.IV: 13989081 - 14012074 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268846.3 NP_001255775.1 12 - 13 1895 79
NM_001268847.4 NP_001255776.1 11 - 12 1744 79
NM_001307070.3 NP_001293999.1 11 - 12 1717 79

View Full Product Details