Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
hhat-2
Entrez Gene ID
190374
Gene Name
Homolog of Hedgehog AcylTransferase
Gene Aliases
CELE_Y57G11C.17
Chromosome Location
Chr.IV: 14826028 - 14829126 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268911.3 NP_001255840.1 4 - 5 930 88

View Full Product Details