Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srt-48
Entrez Gene ID
189835
Gene Name
Serpentine Receptor, class T
Gene Aliases
CELE_Y41E3.15
Chromosome Location
Chr.IV: 15065866 - 15068918 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070420.3 NP_502821.3 3 - 4 709 121

View Full Product Details