Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mel-46
Entrez Gene ID
178469
Gene Name
protein-coding
Gene Aliases
CELE_T06A10.1
Chromosome Location
Chr.IV: 16855044 - 16863449 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001268980.3 NP_001255909.1 8 - 9 2452 86
NM_001268982.3 NP_001255911.1 3 - 4 1706 86
GenBank mRNA EU051652.1 - 8 - 9 2468 86

View Full Product Details