Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
uaf-2
Entrez Gene ID
178489
Gene Name
protein-coding
Gene Aliases
CELE_Y116A8C.35
Chromosome Location
Chr.IV: 17116771 - 17118767 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_070635.5 NP_503036.1 3 - 4 499 70

View Full Product Details